Baseline Goniometer
Supplier: AMERICAN 3B SCIENTIFIC SE
1007371
470119-604EA 76.15 CAD
470119-604
Baseline Goniometer
Goniometers
This stainless steel goniometer measures the range of motion of finger joints (metacarpophalangeal and interphalangeal) and other small joints including toes.
The goniometer's head has two opposing 180º scales marked in 5º increments. A linear scale in inches and centimeters is on arm of goniometer.