Amplitude™ Lambda™ Wipes
Supplier: Contec
AMLA0002 AMLA0011
76470-816CS 500.62 CAD
76470-816 76945-150 76945-152 76945-154
Amplitude™ Lambda™ Wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Amplitude™ Lambda™ Wipes are a hydroentangled blend of 58% cellulose and 42% polyester. The Lambda™ wipe is highly sorbent, has excellent durability, and good cleanliness.
  • Hydroentangled fabric provides low levels of particles and extractables
  • Excellent for wiping and absorbing spills
  • Strong and durable both wet and dry
  • Good cost effective, general-purpose wipe
An excellent all-purpose wipe with outstanding sorbency, it is ideal for general-purpose cleaning and spill pick-up in biotech, pharmaceutical, and medical device industries.
Recommended for ISO Class 6 – 8 environments.