Basic B Lymphocyte Backbone Panel [clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30]Basic B Lymphocyte Backbone Panel

Supplier: Tonbo Biosciences
91-0100-T025 91-0100-T100
10002-356EA 759.98 CAD
10002-356 10002-358
Basic B Lymphocyte Backbone Panel [clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30]Basic B Lymphocyte Backbone Panel
Antibodies
The Human Basic B Lymphocyte backbone panel is a six-color combination of anti-human antibodies. This panel can serve as a backbone to identify the various B lymphocyte subsets, including naive, memory and plasma cells. This panel configuration leaves room for GFP or additional drop-in markers, if desired. Violet laser (405 nm) required for detection of CD45 violetFluoTM 450.

Contains:
1 x PerCP-Cyanine5.5 Anti-Human CD4 (OKT4)
1 x PE Anti-Human CD19 (SJ25C1)
1 x redFluor™ 710 Anti-Human CD20 (2H7)
1 x PE-Cyanine7 Anti-Human CD27 (O323)
1 x APC Anti-Human CD38 (HIT2)
1 x violetFluor™ 450 Anti-Human CD45 (HI30)

Type: Primary
Antigen: CD4, CD19, CD20, CD27, CD38, CD45
Clonality: Monoclonal
Clone: OKT4, SJ25C1, 2H7, O323, HIT2, HI30
Conjugation: PerCP-Cy5.5, PE, redFluor® 710, PE-Cy7, APC (Allophycocyanin), violetFluor® 450
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR