Anti-FCER2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: EBVCS-5]

Supplier: Tonbo Biosciences
20-0237-T025 20-0237-T100 20-0237-T500
10002-108EA 181.7 CAD
10002-108 10002-110 10002-112
Anti-FCER2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: EBVCS-5]
Antibodies
The EBVCS-5 antibody is specific for human CD23, also known as the low affinity IgE receptor (FceRII), a 45 kDa type II transmembrane protein and member of the immunoglobulin superfamily. It is expressed on most B cells and is upregulated upon activation. It is also present on mantle zone B cells, eosinophils, monocytes and a subset of T cells and platelets. CD23 plays a role in B cell development and differentiation, and also functions to regulate IgE production. Soluble secreted forms of CD23 have been reported to bind IgE, stimulate release of pro-inflammatory cytokines from monocytes, and play a role in B cell differentiation.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone: EBVCS-5
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR