Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 61D3]

Supplier: Tonbo Biosciences
20-0149-T025 20-0149-T100 20-0149-T500
10753-532EA 118.5 CAD
10753-532 10753-716 10753-718
Anti-CD14 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 61D3]
Antibodies
The 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol CD14
Antigen name CD14 Molecule
Environmentally Preferable
Antigen synonyms monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein
Antibody type Primary
Clonality Monoclonal
Conjugation APC (Allophycocyanin)
Clone 61D3
Reactivity Human
Host Mouse
Gene ID 929
Isotype IgG1 kappa
Flow cytometry Yes
Cross adsorption No
Storage temperature Store undiluted at 2-8°C, protect from light, do not freeze.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR