Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: L17F12]

Supplier: Tonbo Biosciences
20-0058-T025 20-0058-T100 20-0058-T500
10753-526EA 181.7 CAD
10753-526 10753-528 10753-530
Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: L17F12]
Antibodies
The L17F12 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is a member of the scavenger receptor superfamily and is present on approximately 70% of normal peripheral blood lymphocytes. CD5 is involved in modulating antigen receptor signaling in both T and B cells and acts through binding of CD72, its receptor expressed on B cells.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR