Omni Mixer and Macro Homogenizers, Omni International
Supplier: Omni International
17106 17506 17505
CA11011-580EA 15402.95 CAD
CA11011-580 CA11011-584 10015-894
Omni Mixer and Macro Homogenizers, Omni International
Homogenizers Benchtop Homogenizers
Since 1956 the Omni Mixer Homogenizers have been synonymous with laboratory homogenizers.
The Omni Mixer Homogenizer is compatible with stainless steel generator probes ranging in diameter from 7 to 35 mm, and glass, plastic, or stainless steel chambers from 13 to 1360 ml with sealed generator probes or blades. The Omni Macro Homogenizer is compatible with stainless steel generator probes with a 7, 10, 20, 32, 45, and 55 mm diameter, and glass, plastic, or stainless steel chambers from 13 to 1360 ml with sealed generator probes or blades.
The Omni Mixer Homogenizer features aMore Product Information