Syringe Filters with Luer Lock Inlet, Restek
Supplier: Restek
23978 23979 23966 23967 23968 23969 23974 23975 26142 26143 26144 26145 26146 26147 26148 26149
10059-128PK 397.39 CAD
10059-128 10059-130 10057-936 10057-938 10057-940 10057-942 10057-944 10057-946 10059-502 10059-504 10059-506 10059-508 10059-510 10059-512 10059-514 10059-516
Syringe Filters with Luer Lock Inlet, Restek
Filters Syringe Filters
Syrings filters have guaranteed leak-tight connections with luer lock inlets.
  • Variety of filter types, porosities, and diameters
  • Color-coded for easy identification
  • Rugged polypropylene housing
  • Autoclavable to 121 °C for 15 minutes
  • Quantity break pricing for greater savings
  • Cellulose Acetate, Nylon, PES, PVDF - hydrophilic applications
  • PTFE - hydrophobic applications
Syringe filters are for laboratory use only.