Water Soluble Bags, Elkay Plastics
Supplier: Elkay Plastics
WSB3639 WSB2633 WSB2839
10790-158CS 430.68 CAD
10790-158 10790-352 10790-354
Water Soluble Bags, Elkay Plastics
Bags Waste Disposal Bags
These water soluble bags are an environmentally-friendly and hygienic solution to minimize contact between staff and contaminated laundry.
Product protects staff from risk of contamination and dissolves completely in water temperature of 140 °F or above.
Ordering information: Tie tape attached to bag for easy closure. Bags can hold wet or damp laundry for hours and will not damage laundry, equipment or sewer system when properly used.