Ice Bags, Elkay Plastics
Supplier: Elkay Plastics
H18MET H20MET H21MET H28MET
10790-276CS 190.65 CAD
10790-276 10790-278 10790-280 10790-282
Ice Bags, Elkay Plastics
Bags Storage Bags
These Metallocene ice bags are made from a production process that yields a special balance of properties that are ideal for ice bag and cold storage applications.
  • High impact strength
  • Low haze/high gloss and clarity
Certifications: Bags meet FDA requirements for contact with food products.
Order Now
SPECIFICATIONS
Environmentally Preferable