Clean Scrub™ Mop, Contec®
Supplier: Contec
VKSC0003 VKSC0004
10017-772CS 812.49 CAD
10017-772 10811-978
Clean Scrub™ Mop, Contec®
Mops
Contec’s Clean Scrub™ single-use mop head is wrapped with a textured polyester mesh to eliminate dirt and grime from heavily contaminated surfaces. The mop head has a core of polyester fabric and foam that provides exceptional liquid absorption and resists tearing.
  • Textured polyester mesh removes heavily soiled contamination from surfaces
  • Single-use mop head eliminates cross contamination
  • Slim profile fits into small spaces
  • Compatible with a wide range of disinfectants
This pad is fastened to a semi-rigid thermoformed plate that easily snaps ontoMore Product Information