Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Supplier: Diversified Biotech
GRDT-2000 GRDT-4000
10141-768RL 143.87 CAD
10141-768 10141-770 10141-772
Outdoor Labels for Moist, Frosty or Frozen Surfaces, Diversified Biotech
Labels
Grips to wet, frost-covered and frozen surfaces.
  • Thin material is compatible with centrifuge rotors
  • Resistant to chemicals and solvents
  • Adheres to plastics, glass, metals and even paperboard boxes
  • Temperature range is –80 to 70 °C
Choose your format: laser sheets for printing many labels at once, direct thermal labels for printing a few labels at a time or labels on a roll for handwriting labels.