Lables, Diversified Biotech
Supplier: Diversified Biotech
Direct Thermal Cryo-Tags®
DTCR-6000-R DTCR-6000-O DTCR-6000-Y DTCR-6000-B DTCR-6000-G
490006-742EA 200.72 CAD
490006-742 10141-654 10141-660 10141-658 10141-656
Lables, Diversified Biotech
Labels
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers
Quickly label samples with sharp text, clear barcodes or a combination of both. For use in Temperatures From 70 to –196 °C.