Cryo Labels, Diversified Biotech
Supplier: Diversified Biotech
DTCR-4000-B DTCR-4000-G DTCR-4000-O DTCR-4000-R DTCR-4000-Y
10141-644RL 191.31 CAD
10141-644 10141-646 10141-648 10141-650 10141-652
Cryo Labels, Diversified Biotech
Labels
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers.
Quickly label samples with sharp text, clear barcodes or a combination of both. For use in Temperatures From 70 to –196 °C.