Cryo Labels, Diversified Biotech
Supplier: Diversified Biotech
DTCR-3000-Y DTCR-3000-B DTCR-3000-G DTCR-3000-O DTCR-3000-R
490006-734EA 152.96 CAD
490006-734 10141-636 10141-638 10141-640 10141-642
Cryo Labels, Diversified Biotech
Labels
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers.
Quickly label samples with sharp text, clear barcodes, or a combination of both. For use in temperatures from 70 to –196 °C.