Cryo Labels, Diversified Biotech
Supplier: Diversified Biotech
DTCR-2000-B DTCR-2000-G DTCR-2000-O DTCR-2000-R DTCR-2000-Y
10141-626RL 132.83 CAD
10141-626 10141-628 10141-630 10141-632 10141-634
Cryo Labels, Diversified Biotech
Labels
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers.
Quickly label samples with sharp text, clear barcodes, or a combination of both. For use in temperatures from 70 to –196 °C.