Cryo Labels, Diversified Biotech
Supplier: Diversified Biotech
DTCR-1000-B DTCR-1000-G DTCR-1000-O DTCR-1000-R DTCR-1000-Y
10141-944RL 133.51 CAD
10141-944 10141-946 10141-948 10141-950 10141-956
Cryo Labels, Diversified Biotech
Labels
Direct Thermal Cryo-Tags® are formatted to easily work with the Dymo® Labelwriter®, but also with most direct thermal printers.
Quickly label samples with sharp text, clear barcodes, or a combination of both. For use in temperatures from 70 to –196 °C.