Cryo Labels, Laser Printable, Diversified Biotech
Supplier: Diversified Biotech
LCRY-1100-B LCRY-1100-G LCRY-1100-O LCRY-1100-R LCRY-1100-Y
10141-608PK 178.02 CAD
10141-608 10141-610 10141-612 10141-614 10141-616
Cryo Labels, Laser Printable, Diversified Biotech
Labels
Cryo-Babies® and Cryo-Tags® withstand both long term cryogenic storage conditions and conventional freezer storage
  • Ideal use for Cryovials
  • Temperature ranges from –196 to 150 °C
These labels are chemically inert with a solvent resistant adhesive that will adhere to cryogenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Babies® and Cryo-Tags® also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100 °C and dry heat up to 150 °C (see temp. ranges).