Lab Seal™ Tamper-Evident Specimen Bag, Elkay Plastics
Supplier: Elkay Plastics
LABA610GR LABA610YE LABA610PU LABA610RST LABA610YST LABA610ABS LABA610 LABA1010B LABA1010 LABA1318B LABA610B LABA610BL LABAC20610 LABAC20610STAT LABAC21010 LABAC21517 LABAC220606 LABAC221318
10789-522CS 332.26 CAD
10789-522 10789-524 10789-526 10789-528 10789-530 10789-532 10789-534 10789-536 10789-538 10789-540 10789-662 10789-664 10790-104 10790-106 10790-108 10790-110 10790-112 10790-114
Lab Seal™ Tamper-Evident Specimen Bag, Elkay Plastics
Bags Sample Bags
Elkay’s Lab-Seal™ Tamper-Evident specimen bags with Seal-N-Rip™ removable biohazard symbol provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
  • A special mechanism where once its marked tab is pulled, the bag will seal securely and permanently with adhesive glue
Once the bag is sealed, the only way to access the contents would be to tear off the top indicated by a tear line, which would reveal any tampering.
The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncontaminated.