Lab Loc® Specimen Bags, Elkay Plastics
Supplier: Elkay Plastics
LABZ69BBX LABZ69BL LABZ69YE LABZ69PU LABZ69OR LABZ69RE LABZ69BA LABZ810B LABZ69RST LABZ69YST LABZ46B LABZ69B LABZ69
10789-494CS 217.17 CAD
10789-494 10789-496 10789-500 10789-502 10789-504 10789-506 10789-508 10789-510 10789-632 10789-634 10790-168 10790-170 10026-352
Lab Loc® Specimen Bags, Elkay Plastics
Bags Sample Bags
Elkay’s Lab-Loc® reclosable seal top specimen bags with Seal-N-Rip™ removable biohazard symbols provide an means for securely transferring specimens in hospital and medical environments while helping to save on disposal costs.
Printed in both English and Spanish and available in a variety of sizes, colors and printed messages for various types of specimens.
The bags have a special removable biohazard symbol which can be torn off and allows the bags to be disposed of with regular waste if they are uncominated. Their three wall construction provides a handy outer pouch to contain accompanying paperwork during specimen transfer.
Packaging: Bags are inner-packed in individually-marked and barcoded dispenser packs for convenience.