ContainerSeal™, Tape, Diversified Biotech
Supplier: Diversified Biotech
CSEAL-75R CSEAL-75W CSEAL-75Y CSEAL-XLW
10141-786RL 117.46 CAD
10141-786 10141-788 10141-790 10141-794
ContainerSeal™, Tape, Diversified Biotech
Tapes
Sealing tape for lids, caps, and other lab items.
  • Stretches, conforms, and seals the perimeter of microplates and petri dishes
  • Waterproof, resists most solvents and caustic agents
  • Recommended temperature range: -40 to 37 °C
Seal remains flexible in moderate freezer temperatures and withstands intermittent exposure to 100 °C.