Head Mirror, Sklar
Supplier: Sklar
73-1015 73-1086
10198-592EA 328.31 CAD
10198-592 10198-596
Head Mirror, Sklar
Mirrors
The Head Mirror is used in conjunction with a headband (sold seperately) as a diagnostic tool, often as part of an ENT examination
The round mirror features a small center hole. It is situated on the users forehead via the adjustable length headband. The mirror is concave and concentrates light onto specific areas without shadows. Each mirror also features a black, non-latex, protective ring around the exterior.