Titanium(IV) butanolate 99+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Titanium (IV) butoxide, Titanium tetrabutanolate, Tetra-n-butyl orthotitanate, Titanium (IV) n-butoxide
- Liquid
Purity: 99+%
CAS No.: 5593-70-4
Molecular Formula: C16H36O4Ti
Formula Weight: 340.36
Flash Point: 41°(170°F)
Physical Form: Liquid
Density: 0.998
Refractive Index: 1.4900
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
Odor: Alcohol-like
Melting Point: -55°
Boiling Point: 310-314°
Solubility: Soluble in organic solvents. Decomposes in water
Beilstein Reference: 4148236
UN No.: 2924
MDL No.: MFCD00009433
Formula:
C₁₆H₃₆O₄Ti MW: 340.34 g/mol Boiling Pt: 310 Melting Pt: -55 Density: 0.998 Flash Pt: 41 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009433 CAS Number: 5593-70-4 EINECS: 227-006-8 UN: 2924 ADR: 3 III |
Specification Test Results
Assay Minimum | 99 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...