Aluminium oxide ≥99%

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

047171-36 047171-A3
AA47171-36EA 145.23 CAD
AA47171-36 AA47171-A3
Aluminium oxide ≥99%
Aluminium oxide

32-63 Micron APS Powder, S.A. 200m2/g


Purity: 99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 32-63 Micron APS Powder, S.A. 200m2/g Density: 3.965 Refractive Index: 1.768 Appearance: White Odor: Odorless Melting Point: 2045° Boiling Point: 2980° MDL No.: MFCD00003424

Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2980
Melting Pt: 2045
Density: 3.965
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR