Platinum, expanded metal mesh, gauze, Unimesh N7433, Thickness: 0.34 mm (0.013 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

046966-FF 046966-FI 46966-FF 46966-FI
CAAA46966-FFEA 666 CAD
AA46966-FI CAAA46966-FF CAAA46966-FI AA46966-FF
Platinum, expanded metal mesh, gauze, Unimesh N7433, Thickness: 0.34 mm (0.013 in)
Platinum

  • ~1.0g/25x25mm, Mesh

Formula: Pt
MW: 195.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR