Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
46943-GH
46943-HW
46943-HX
CAAA46943-GHEA
113.92
CAD
CAAA46943-GH
CAAA46943-HW
CAAA46943-HX
Molybdenum ≥99.95% (metals basis), foil, Thickness: 0.05 mm (0.002 in)
Molybdenum
- ~5.1g/100x100mm
Purity: 99.95%
CAS No.: 7439-98-7
Physical Form: ≈5.1g/100x100mm
MDL No.: MFCD00003465
Formula:
Mo Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 |
Specification Test Results
Assay Minimum | 99.95 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...