Potassium peroxodisulfate ≥99.0% ACS low in nitrogen

Supplier: Thermo Scientific Chemicals
Danger

46939-30 46939-A1
CAAA46939-30EA 96.54 CAD
CAAA46939-30 CAAA46939-A1
Potassium peroxodisulfate ≥99.0% ACS low in nitrogen
Potassium peroxodisulphate

  • Crystalline


Purity: 99.0% min CAS No.: 7727-21-1 Molecular Formula: K2O8S2 Formula Weight: 270.31 Physical Form: Crystalline Density: 2.477 Appearance: White Odor: Odorless Melting Point: 100° dec. UN No.: 1492 MDL No.: MFCD00011386

Formula: K₂S₂O₈
Melting Pt: 100 °C (dec.)
Density: 2.477
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 14,7656

Order Now

Specification Test Results

Assay Minimum 99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR