Sodium fluoride ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

46912-09 46912-18
CAAA46912-09EA 93.38 CAD
CAAA46912-09 CAAA46912-18
Sodium fluoride ≥99.99% (metals basis)
Sodium fluoride

  • 3-6mm pieces


Purity: 99.99% CAS No.: 7681-49-4 Molecular Formula: FNa Formula Weight: 41.99 Physical Form: 3-6mm pieces Density: 2.78 Refractive Index: 1.336 Appearance: White Odor: Odorless Melting Point: 993° Boiling Point: 1704° UN No.: 1690 MDL No.: MFCD00003524

Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1704 °C
Melting Pt: 993 °C
Density: 2.78
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1,III
Merck Index: 14,8618
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Minimum 99.99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR