Bismuth indium tin, ingot
Supplier: Thermo Scientific Chemicals
Synonyms:
Field's metal
46895-22
46895-36
CAAA46895-22EA
336.54
CAD
CAAA46895-22
CAAA46895-36
CAAA46895-A3
Bismuth indium tin, ingot
Bismuth indium tin
Ingot
Molecular Formula: Bi:In:Sn; 32.5:51:16.5 wt%
Physical Form: Ingot
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...