1-Hydroxytetraphenylcylclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II) 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Shvo's Catalyst, 1-Hydroxytetraphenylcyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-mu-hydrotetracarbonyldiruthenium(II), SHVO'S CATALYST, 1-Hydroxytetraphenyl-cyclopentadienyl(tetraphenyl-2,4-cyclopentadien-1-one)-u-hydrotetracarbonyldiruthenium(II)
- Crystalline
Purity: 98%
CAS No.: 104439-77-2
Molecular Formula: C62H42O6Ru2
Formula Weight: 1085.15
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Appearance: Yellow to orange
Melting Point: 223-227°
MDL No.: MFCD03840556
Applications: Calayst for hydrogen transfers including reduction of ketones and aldehydes, and oxidation and/or racemization of amines and alcohols
Formula:
C₆₂H₄₂O₆Ru₂ MW: 1085.15 g/mol Melting Pt: 223…227 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840556 CAS Number: 104439-77-2 Merck Index: 14,8483 |
Specification Test Results
Assay Minimum | 98 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...