Dicyclopentenyl acrylate
Supplier: Thermo Scientific Chemicals
|
Danger
|
46864-14
46864-22
CAAA46864-14EA
138.57
CAD
CAAA46864-14
CAAA46864-22
Dicyclopentenyl acrylate
Dicyclopentenyl acrylate
- Liquid
CAS No.: 33791-58-1
Molecular Formula: C13H16O2
Formula Weight: 204.27
Storage Temperature: 2°C to 8°C
Flash Point: 131°(268°F)
Physical Form: Liquid
Density: 1.07
Appearance: Pale brown
Melting Point: -40°
MDL No.: MFCD00171305
Formula:
C₁₃H₁₆O₂ Melting Pt: –40 °C Density: 1.07 Flash Pt: 131 °C (268 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00171305 CAS Number: 33791-58-1 EINECS: 251-678-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...