Copper, gauze woven from wire 30 mesh Ø 0.15 mm (0.006 in)
Supplier: Thermo Scientific Chemicals
46834-FL
46834-GT
46834-HB
CAAA46834-FLEA
44.87
CAD
CAAA46834-FL
CAAA46834-GT
CAAA46834-HB
Copper, gauze woven from wire 30 mesh Ø 0.15 mm (0.006 in)
Copper
- Wire Cloth
CAS No.: 7440-50-8
Physical Form: Wire Cloth
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2595 Melting Pt: 1083 Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...