Titanium, gauze woven from wire 60 mesh Ø 0.2 mm (0.009 in)

Supplier: Thermo Scientific Chemicals

46828-FL 46828-GT 46828-HB
CAAA46828-FLEA 113.13 CAD
CAAA46828-FL CAAA46828-GT CAAA46828-HB
Titanium, gauze woven from wire 60 mesh Ø 0.2 mm (0.009 in)
Titanium

  • Wire Cloth


CAS No.: 7440-32-6 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR