Titanium, gauze woven from wire 80 mesh Ø 0.13 mm (0.005 in)

Supplier: Thermo Scientific Chemicals

46799-FL 46799-GT
CAAA46799-FLEA 143.46 CAD
CAAA46799-FL CAAA46799-GT
Titanium, gauze woven from wire 80 mesh Ø 0.13 mm (0.005 in)
Titanium

  • Wire Cloth


CAS No.: 7440-32-6 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR