Inconel® 600 alloy, gauze woven from wire 200 mesh Ø 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
46791-FL
46791-GT
CAAA46791-FLEA
83.74
CAD
CAAA46791-FL
CAAA46791-GT
Inconel® 600 alloy, gauze woven from wire 200 mesh Ø 0.05 mm (0.002 in)
Inconel® 600 alloy
Wire Cloth
Physical Form: Wire Cloth
MDL No.: MFCD01741927
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...