Tetrabutylammonium dihydrogen phosphate ≥98%, non-UV
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetra-n-butylammonium phosphate monobasic, Tetrabutylammonium phosphate monobasic, Tetra-n-butylammonium dihydrogen phosphate, IPC-TBA-P, Tetra-n-butylammonium phosphate
Tetra-n-butylammonium phosphate, is used in chemicals, pharmaceutical research, pharmaceutical intermediates, and reagents. Tetrabutylammonium phosphate can be used in organic synthesis. It is used as an Ion-associating reagent for HPLC.
Purity: 98%
CAS No.: 5574-97-0
Molecular Formula: C16H38NO4P
Formula Weight: 339.46
Physical Form: Crystalline
Appearance: White
Melting Point: 151-154°
Beilstein Reference: 5196532
MDL No.: MFCD00064526
Formula:
C₁₆H₃₈NO₄P Melting Pt: 151…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00064526 CAS Number: 5574-97-0 EINECS: 226-947-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...