Gadolinium nitride ≥99.5% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Gadolinium(III) nitride

46709-03 46709-06
CAAA46709-03EA 53.25 CAD
CAAA46709-03 CAAA46709-06
Gadolinium nitride ≥99.5% (REO, rare earth oxide basis)
Gadolinium nitride

  • -60 Mesh Powder


Purity: 99.5% CAS No.: 25764-15-2 Molecular Formula: GdN Formula Weight: 171.26 Physical Form: -60 Mesh Powder Appearance: Black MDL No.: MFCD00049943

Formula: GdN
MW: 171.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049943
CAS Number: 25764-15-2
EINECS: 247-250-9

Order Now

Specification Test Results

Assay Minimum 99.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR