Hastelloy C® alloy, gauze woven from wire 40 mesh Ø 0.19 mm (0.0075 in)
Supplier: Thermo Scientific Chemicals
46690-FL
46690-GT
46690-HB
CAAA46690-FLEA
61.15
CAD
CAAA46690-FL
CAAA46690-GT
CAAA46690-HB
Hastelloy C® alloy, gauze woven from wire 40 mesh Ø 0.19 mm (0.0075 in)
Hastelloy C® alloy
Wire Cloth
Physical Form: Wire Cloth
MDL No.: MFCD00801065
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...