Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

46661-G1 46661-G5 46661-G9
CAAA46661-G1EA 176.96 CAD
CAAA46661-G1 CAAA46661-G5 CAAA46661-G9
Indium ≥99.998% (metals basis), wire, Puratronic®, annealed, Ø 1.0 mm (0.04 in)
Indium

  • ~5.74g/m


Purity: 99.998+% CAS No.: 7440-74-6 Physical Form: ≈5.74g/m Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now

Specification Test Results

Assay Minimum 99.998


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR