Copper, gauze woven from wire 140 mesh Ø 0.056 mm (0.0022 in)

Supplier: Thermo Scientific Chemicals

46603-FL 46603-GT 46603-HB
CAAA46603-FLEA 48.66 CAD
CAAA46603-FL CAAA46603-GT CAAA46603-HB
Copper, gauze woven from wire 140 mesh Ø 0.056 mm (0.0022 in)
Copper

  • Wire Cloth


CAS No.: 7440-50-8 Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR