3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

Supplier: Thermo Scientific Chemicals
Danger

46539-06 46539-14 46539-22
CAAA46539-06EA 50.56 CAD
CAAA46539-06 CAAA46539-14 CAAA46539-22
3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate
3-(2,2,3-Trimethylnorborn-5-yl)cyclohexyl acrylate

  • Liquid


CAS No.: 903876-45-9 Molecular Formula: C19H30O2 Formula Weight: 290.45 Storage Temperature: 2°C to 8°C Flash Point: 149°(300°F) Physical Form: Liquid Appearance: Pale brown MDL No.: MFCD22380703

Formula: C₁₉H₃₀O₂
Flash Pt: >149 °C (300 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD22380703
CAS Number: 903876-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR