Tungsten, gauze woven from wire 40 mesh woven from 0.025mm (0.001in)

Supplier: Thermo Scientific Chemicals

46412-FL 46412-GT
CAAA46412-FLEA 80.26 CAD
CAAA46412-FL CAAA46412-GT
Tungsten, gauze woven from wire 40 mesh woven from 0.025mm (0.001in)
Tungsten

  • Wire Cloth


CAS No.: 7440-33-7 Physical Form: Wire Cloth MDL No.: MFCD00011461

Formula: W
MW: 183.84 g/mol
Boiling Pt: 5660
Melting Pt: 3410
Storage Temperature: Ambient
MDL Number: MFCD00011461
CAS Number: 7440-33-7
EINECS: 231-143-9
Merck Index: 12,09945

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR