Titanium, gauze woven from wire Ø 0.05 mm (0.002 in) 100 mesh
Supplier: Thermo Scientific Chemicals
046406-GT
46406-FL
046406-FL
46406-GT
CAAA46406-FLEA
202.24
CAD
CAAA46406-FL
AA46406-FL
CAAA46406-GT
AA46406-GT
Titanium, gauze woven from wire Ø 0.05 mm (0.002 in) 100 mesh
Titanium
Wire Cloth
Formula:
Ti MW: 47.88 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011264 CAS Number: 7440-32-6 EINECS: 231-142-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...