EMCA (6-Maleimidohexanoic acid), powder
Supplier: Thermo Scientific Chemicals
|
Warning
|
46384-06
46384-14
46384-22
CAAA46384-06EA
91.64
CAD
CAAA46384-06
CAAA46384-14
CAAA46384-22
EMCA (6-Maleimidohexanoic acid), powder
EMCA (6-Maleimidohexanoic acid)
CAS No.: 55750-53-3
Molecular Formula: C10H13NO4
Formula Weight: 211.22
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to pale yellow
MDL No.: MFCD00043140
Formula:
C₁₀H₁₃NO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00043140 CAS Number: 55750-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...