Strontium peroxide (∼12.3% O₂)
Supplier: Thermo Scientific Chemicals
|
Danger
|
46310-30
46310-A1
46310-A7
CAAA46310-30EA
111.86
CAD
CAAA46310-30
CAAA46310-A1
CAAA46310-A7
Strontium peroxide (∼12.3% O₂)
Strontium peroxide
CAS No.: 1314-18-7
Molecular Formula: O2Sr
Formula Weight: 119.62
Physical Form: -100 Mesh Powder
Sensitivity: Moisture Sensitive
Appearance: Pale brown
Melting Point: 215° dec.
UN No.: 1509
MDL No.: MFCD00084686
Product Specifications are subject to amendment and may change over time.
Formula:
O₂Sr Melting Pt: 215 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00084686 CAS Number: 1314-18-7 EINECS: 215-224-6 UN: 1509 ADR: 5.1,II Merck Index: 14,8847 |
Specification Test Results
Available Oxygen | 12.3% min |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...