Copper ≥99.99% (metals basis), thin foil 0.008 mm (0.0003 in) thick, Oxygen-Free High Conductivity (OFHC)

Supplier: Thermo Scientific Chemicals

45943-FF 45943-FI 45943-GH
CAAA45943-FFEA 92.59 CAD
CAAA45943-FF CAAA45943-FI CAAA45943-GH
Copper ≥99.99% (metals basis), thin foil 0.008 mm (0.0003 in) thick, Oxygen-Free High Conductivity (OFHC)
Copper

  • ~0.04g/25x25mm


Purity: 99.99% CAS No.: 7440-50-8 Physical Form: ≈0.04g/25x25mm Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

Specification Test Results

Assay Minimum 99.99


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR