Palladium 10% on activated carbon, ca. 50% moisture, powder, reduced Type 87L

Supplier: Thermo Scientific Chemicals

45924-06 45924-14
CAAA45924-06EA 260.7 CAD
CAAA45924-06 CAAA45924-14
Palladium 10% on activated carbon, ca. 50% moisture, powder, reduced Type 87L
Palladium

  • 900-1100m2/g


CAS No.: 7440-05-3 Physical Form: 900-1100m2/g Appearance: Black MDL No.: MFCD03457879

Formula: Pd
Storage Temperature: Ambient
MDL Number: MFCD03457879
CAS Number: 7440-05-3
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR