Lithium perchlorate trihydrate ACS

Supplier: Thermo Scientific Chemicals
Danger

45923-22 45923-36
CAAA45923-22EA 115.02 CAD
CAAA45923-22 CAAA45923-36
Lithium perchlorate trihydrate ACS
Lithium perchlorate trihydrate

Lithium perchlorate trihydrate is used as an oxygen source for chemical oxygen generators as it liberates oxygen at high temperatures. Excellent solubility of lithium perchlorate permits its use in diels-alder reactions.


Lithium perchlorate trihydrate is also employed in lithium-ion batteries as an electrolyte and acts as a chaotropic agent to denature proteins. It is used in the baylis-hillman reaction as a catalyst in the coupling of alpha, beta-unsaturated carbonyl compounds with aldehydes.

Formula: ClLiO₄·3H₂O
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00149765
CAS Number: 13453-78-6
EINECS: 232-237-2
UN: 1481
ADR: 5.1,II
Merck Index: 14,5539

Order Now

Specification Test Results

Assay (Unspecified) 63.0 - 68.0% LiClO₄
Heavy Metals (ICP-OES) 5 ppm
Impurity Content, Cl 0.003%
Impurity Content, Fe 5 ppm
Impurity Content, SO₄ 0.001%
Insoluble Material 0.005%
pH 6.0 - 7.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR