Holmium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

45919-06 45919-14
CAAA45919-06EA 246.48 CAD
CAAA45919-06 CAAA45919-14
Holmium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Holmium

  • Packaged under argon


Purity: 99.9% CAS No.: 7440-60-0 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00011049

Formula: Ho
Storage Temperature: Ambient
MDL Number: MFCD00011049
CAS Number: 7440-60-0
EINECS: 231-169-0
UN: 3089
ADR: 4.1,II

Order Now

Specification Test Results

Assay Minimum 99.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR