Dysprosium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

45918-06 45918-14
CAAA45918-06EA 589.34 CAD
CAAA45918-06 CAAA45918-14
Dysprosium ≥99.9% (REO, rare earth oxide basis), powder -200 mesh
Dysprosium

  • Packaged under argon


Purity: 99.9% CAS No.: 7429-91-6 Physical Form: Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Silver Odor: Odorless UN No.: 3089 MDL No.: MFCD00010982

Formula: Dy
MW: 162.5
Storage Temperature: Ambient
MDL Number: MFCD00010982
CAS Number: 7429-91-6
EINECS: 231-073-9
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Assay Minimum 99.9


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR